Merck
  • One-step preparation of S-nitrosated human serum albumin with high biological activities.
One-step preparation of S-nitrosated human serum albumin with high biological activities.
Nitric oxide : biology and chemistry (2010-05-11)
Yu Ishima, Shuichi Hiroyama, Ulrich Kragh-Hansen, Toru Maruyama, Tomohiro Sawa, Takaaki Akaike, Toshiya Kai, Masaki Otagiri
ABSTRACT
S-Nitrosated human serum albumin (SNO-HSA) is a large molecular weight nitric oxide carrier in human plasma, and because of its many beneficial effects in different tests, it is currently under investigation as a cytoprotective agent. However, making SNO-HSA preparations is a complicated and time-consuming process. We found that binding of caprylic acid (CA) and N-acetyl-l-tryptophan (N-AcTrp) to defatted mercaptalbumin increased S-nitrosation by S-nitrosoglutathione (GS-NO) by making Cys-34 of HSA more accessible and by protecting it against oxidation, respectively. Fortunately, HSA solutions for clinical use contain high concentrations of CA and N-AcTrp as stabilizers. By making use of that fact it was possible to work-out a fast and simple procedure for producing SNO-HSA: incubation of a commercial HSA formulation with GS-NO for only 1 min results in S-nitrosation of HSA. The biological usefulness of such a preparation was tested in a rat ischemia-reperfusion liver injury model. Although our procedure for making SNO-HSA is fast and straightforward, the cytoprotective effect of the preparation was similar to, or better than, that of a preparation made in a more traditional way. The clinical development of SNO-HSA as a strong cytoprotective agent is under way using this method in collaboration with clinicians and industrial developers.
Sigma-Aldrich
N-Acetyl-L-tryptophan
Supelco
N-Acetyl-L-tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material