Merck
  • Toll-like receptor ligands induce polymorphonuclear leukocyte migration: key roles for leukotriene B4 and platelet-activating factor.
Toll-like receptor ligands induce polymorphonuclear leukocyte migration: key roles for leukotriene B4 and platelet-activating factor.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2009-10-22)
Julie S Lefebvre, Sylvie Marleau, Valérie Milot, Tania Lévesque, Serge Picard, Nicolas Flamand, Pierre Borgeat
ABSTRACT
Activation of toll-like receptors (TLRs) and polymorphonuclear leukocyte (PMN) accumulation at infection sites are critical events of host defense. The involvement of leukotriene (LT) B(4) and platelet-activating factor (PAF) in TLR ligand-induced activation of inflammatory cell functions is essentially unknown. Using an in vitro model of human PMN migration through human endothelial cell monolayers, we demonstrate that prototypic ligands of TLR1/2, 2/6, 3, 4, 5, and 7/8 promote PMN migration, an effect markedly inhibited by 3 LTB(4) receptor antagonists (70-80% inhibition at 100 nM compared to vehicle-treated cells), 3 PAF receptor antagonists (20-50% inhibition at 10 nM), 3 LT biosynthesis inhibitors (75-85% inhibition at 100 nM), and 1 cytosolic phospholipase A(2)alpha (cPLA(2)alpha) inhibitor (90% inhibition at 1 microM). Accordingly, selected TLR ligands caused Ser-505-phosphorylation of cPLA(2)alpha and measurable LTB(4) and PAF biosynthesis in the transmigration assay. As negative controls, interleukin-8- and formyl-methionyl-leucyl-phenylalanine-elicited migration in vitro was not inhibited either by an LTB(4) receptor antagonist or by the cPLA(2)alpha inhibitor. Finally, LTB(4) and PAF receptor antagonists inhibited (up to approximately 65% at optimal doses) TLR ligand-induced PMN infiltration in the mouse air-pouch model. These studies unravel the critical involvement of de novo LTB(4) and PAF biosynthesis in PMN migration elicited by TLR ligands.
Sigma-Aldrich
WEB2086, ≥98% (HPLC)