Merck
  • Epinastine: topical ophthalmic second generation antihistamine without significant systemic side effects.
Epinastine: topical ophthalmic second generation antihistamine without significant systemic side effects.
Expert opinion on drug metabolism & toxicology (2009-07-28)
Shilpi Pradhan, Kumar Abhishek, Francis Mah
ABSTRACT
Epinastine is a topical ophthalmic antihistamine and a mast cell stabilizer indicated for treatment of symptoms associated with allergic conjunctivitis. The objective of this review is to provide a comprehensive overview of the pharmacology, clinical efficacy, tolerability and safety of epinastine 0.05% ophthalmic solution. Medline literature review from 1950 to 2008. Epinastine 0.05% ophthalmic solution was found to be a safe and effective medication for the relief of symptoms of allergic conjunctivitis. Epinastine demonstrates very low CNS and cardiac toxicity.
Sigma-Aldrich
Epinastine hydrochloride, ≥98% (HPLC), solid
Epinastine hydrochloride, European Pharmacopoeia (EP) Reference Standard