Merck
  • Tautomycin suppresses growth and neuroendocrine hormone markers in carcinoid cells through activation of the Raf-1 pathway.
Tautomycin suppresses growth and neuroendocrine hormone markers in carcinoid cells through activation of the Raf-1 pathway.
American journal of surgery (2009-02-28)
Scott N Pinchot, Joel T Adler, Yinggang Luo, Jianhua Ju, Wenli Li, Ben Shen, Muthusamy Kunnimalaiyaan, Herbert Chen
ABSTRACT
Carcinoids are neuroendocrine (NE) tumors with limited treatment options. Raf-1 pathway activation has been shown to suppress hormone production in carcinoid cells. We investigated a novel treatment for carcinoid cell growth based on pharmacologic Raf-1 activation using the compound tautomycin (TTY). Human carcinoid cells were treated with TTY for 48 hours. Western blot analysis was used to demonstrate Raf-1 pathway activation by phosphorylation of ERK1/2 and to determine the effect on NE tumor markers. Cellular growth was measured by methylthiazolyldiphenyl-tetrazolium bromide (MTT) assay. Treatment with TTY resulted in dose-dependent activation of the Raf-1 pathway. Furthermore, a significant decrease in NE tumor markers was seen. Importantly, TTY inhibited carcinoid cellular growth and induced the cell-cycle inhibitors p21 and p27. TTY activates the Raf-1 pathway, limits carcinoid cell growth, and suppresses NE marker production in vitro. This new compound warrants further investigation in animal models of carcinoid cancer.