Merck
  • Asymmetric synthesis of bicyclo[4.3.1]decadienes and bicyclo[3.3.2]decadienes via [6 + 3] trimethylenemethane cycloaddition with tropones.
Asymmetric synthesis of bicyclo[4.3.1]decadienes and bicyclo[3.3.2]decadienes via [6 + 3] trimethylenemethane cycloaddition with tropones.
Journal of the American Chemical Society (2008-10-22)
Barry M Trost, Patrick J McDougall, Olaf Hartmann, Peter T Wathen
ABSTRACT
The cyanosubstituted trimethylenemethane donor undergoes palladium-catalyzed [6 + 3] cycloaddition with a variety of tropones to yield bicyclo[4.3.1]decadienes in excellent regio-, diastereo-, and enantioselectivity. Products of the Pd-TMM [6 + 3] cycloaddition participate in a thermal [3,3] sigmatropic rearrangement to yield bicyclo[3.3.2]decadienes in good yield.
Sigma-Aldrich
Tropone, 97%