Merck
  • The phospholipidosis-lnducing potential of the chemopotentiating drug, N,N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine (DPPE, tesmilifene) correlates with its stimulation of phosphatidylserine synthesis and exposure on the plasma membrane in MCF-7 breast cancer cells.
The phospholipidosis-lnducing potential of the chemopotentiating drug, N,N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine (DPPE, tesmilifene) correlates with its stimulation of phosphatidylserine synthesis and exposure on the plasma membrane in MCF-7 breast cancer cells.
Proceedings of the Western Pharmacology Society (2008-07-09)
Fred Y Xu, Gary Queen, Lorne Brandes, Grant M Hatch
ABSTRACT
N,N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine (DPPE, tesmilifene), a potent chemopotentiating drug currently in Phase III clinical trials of metastatic breast cancer, increases cytotoxicity of anthracyclines and taxanes in a variety of multi-drug resistance expressing (MDR+) tumor cell lines in vitro; inhibits binding of histamine to CYP3A4, a lipid/prostanoid-regulatory P450; and modulates serum levels of HDL/LDL cholesterol and phospholipids in vivo. Since increased exposure of phosphatidylserine (PS) on the outer cell membrane leaflet is associated with apoptosis, increased clearance of dead cells by phagocytes and inhibition of the P-glycoprotein pump, the effect of DPPE on PS synthesis was assessed in vitro in a human breast cancer cell line. MCF-7 cells were incubated with 5 microM DPPE for 24 hr or 5 days, followed by addition of [1-(14)C]arachidonic acid for 4 hr; or [3H]serine for 8 hr. Compared to untreated cells, a 27-42% (p < 0.05) increase in [1-(14)C]arachidonic acid incorporated into all phospholipids, including a 1.9-fold increase (p < 0.05) in PS was observed in DPPE-treated cells. [3H]Serine incorporation into PS was elevated 37%, while the pool size of PS was elevated 23% (p < 0.05) in DPPE-treated cells, indicating elevated de novo PS biosynthesis. Annexin-5 binding studies indicated an elevation in exposure of PS on the surface of the plasma membrane in DPPE-treated cells. DPPE-treatment also resulted in N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine accumulation indicative of phospholipidosis-inducing potential. Thus, the chemopotentiating effect of DPPE may be due to its phospholipidosis-inducing potential and stimulation of PS synthesis leading to an increased exposure of PS on the cell surface which could potentially enhance cancer cell clearance by phagocytes.
Sigma-Aldrich
DPPE hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.