- [The effect of orthodontic magnets on the oral microbial flora].
[The effect of orthodontic magnets on the oral microbial flora].
Minerva stomatologica (1991-07-01)
N Staffolani, M Cervini, C Giannoni, M Guerra, M Pugliese, F D'Alò, S Farinelli
PMID1753928
ABSTRACT
In this study we wanted to test the in vitro effects of orthodontic magnetic brackets, developing different magnetic fields, on the oral microbial flora. We noticed that a magnetic field has its most considerable influence on Candida albicans growth; the stimulating response depends on various factors: cell inoculum, exposure time and magnetic field frequency.