Merck
  • False susceptibility to cefotetan reported by MicroScan for DHA-type AmpC beta-lactamase-producing Klebsiella pneumoniae.
False susceptibility to cefotetan reported by MicroScan for DHA-type AmpC beta-lactamase-producing Klebsiella pneumoniae.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases (2007-03-03)
S Y Lee, Y J Park, E J Oh, J K Yoo, J J Park, K G Park, K Han
ABSTRACT
This study evaluated the accuracy of cefotetan susceptibility determination using the MicroScan WalkAway system for AmpC-producing Klebsiella pneumoniae. In total, 57 K. pneumoniae isolates that showed a D-shape flattening in a double-disk synergy test were studied. Cefotetan MICs were determined by the agar dilution method. The bla(DHA) gene was detected in all 57 isolates, one of which co-harboured bla(CMY-1). According to the MicroScan system, 28 isolates were susceptible, 18 were intermediately-resistant, and 11 were resistant to cefotetan. Compared with the agar dilution method, very major, minor and major error rates were 28.1% (16/57), 47.4% (27/57) and 1.8% (1/57), respectively.