Merck
  • Combined optimization using Cultural and Differential Evolution: application to crystal structure solution from powder diffraction data.
Combined optimization using Cultural and Differential Evolution: application to crystal structure solution from powder diffraction data.
Chemical communications (Cambridge, England) (2006-10-07)
Samantha Y Chong, Maryjane Tremayne
ABSTRACT
The principles of social and biological evolution have been combined in a Cultural Differential Evolution hybrid global optimization technique and applied to crystal structure solution.