Merck
  • M1 muscarinic receptors contribute to, whereas M4 receptors inhibit, dopamine D1 receptor-induced [3H]-cyclic AMP accumulation in rat striatal slices.
M1 muscarinic receptors contribute to, whereas M4 receptors inhibit, dopamine D1 receptor-induced [3H]-cyclic AMP accumulation in rat striatal slices.
Neurochemical research (2006-06-08)
Enrique Sánchez-Lemus, José-Antonio Arias-Montaño
ABSTRACT
In rat striatal slices labelled with [(3)H]-adenine and in the presence of 1 mM 3-isobutyl-1-methylxantine (IBMX), cyclic [(3)H]-AMP ([(3)H]-cAMP) accumulation induced by the dopamine D(1) receptor agonist SKF-81297 (1 microM; 177 +/- 13% of basal) was inhibited by the general muscarinic agonist carbachol (maximum inhibition 72 +/- 3%, IC(50) 0.30 +/- 0.06 microM). The muscarinic toxin 7 (MT-7), a selective antagonist at muscarinic M(1) receptors, reduced the effect of SKF-81297 by 40+/-7% (IC(50) 251+/- 57 pM) and enhanced the inhibitory action of a submaximal (1 microM) concentration of carbachol (69 +/- 4% vs. 40 +/- 7% inhibition, IC(50) 386 +/- 105 pM). The toxin MT-1, agonist at M(1) receptors, stimulated [(3)H]-cAMP accumulation in a modest but significant manner (137 +/- 11% of basal at 400 nM), an action additive to that of D(1) receptor activation and blocked by MT-7 (10 nM). The effects of MT-7 on D(1) receptor-induced [(3)H]-cAMP accumulation and the carbachol inhibition were mimicked by the PKC inhibitors Ro-318220 (200 nM) and Gö-6976 (200 nM). Taken together our results indicate that in addition to the inhibitory role of M(4) receptors, in rat striatum acetylcholine stimulates cAMP formation through the activation of M(1 )receptors and PKC stimulation.