- Use of omeprazole as a CYP3A probe drug: effect of sex and menstrual cycle phase on CYP3A activity in healthy Caucasian adults.
Use of omeprazole as a CYP3A probe drug: effect of sex and menstrual cycle phase on CYP3A activity in healthy Caucasian adults.
To determine the effects of sex and menstrual cycle phase on CYP3A activity and to characterize the intraindividual variability of CYP3A, 24 Caucasian adults were given a single dose of omeprazole every 14th day for 3 months (men) or during the mid-follicular and mid-luteal phases over 3 full menstrual cycles (women). The 2-hour plasma metabolic ratio (MR) (omeprazole/omeprazole sulphone) was used as a measure of CYP3A activity. Overall mean MRs for men (2.4 +/- 1.2) and women (2.3 +/- 0.93) were not significantly different. In women, no difference in mean omeprazole MR was observed between the mid-follicular phase (2.2 +/- 0.85) and mid-luteal phase (2.4 +/- 1.0). When all data in the mid-luteal phase were stratified into 3 groups based on the progesterone concentrations (>50 nM [A], 20-50 nM [B], and <20 nM [C]), the MR of group A was significantly lower than that of B and C. The MRs of women during the mid-luteal phase were weakly correlated with progesterone concentrations (r = -0.35; P = .03). Individual coefficients of variation (CV%) in MR ranged from 8.7% to 60.2%, with a median 30.0%. No sex or menstrual cycle differences in CYP3A activity as measured by the probe drug omeprazole were seen. Higher CYP3A activity in women may be associated with higher endogenous progesterone concentrations.