- Simultaneous determination of O6-methyl-2'-deoxyguanosine, 8-oxo-7,8-dihydro-2'-deoxyguanosine, and 1,N6-etheno-2'-deoxyadenosine in DNA using on-line sample preparation by HPLC column switching coupled to ESI-MS/MS.
Simultaneous determination of O6-methyl-2'-deoxyguanosine, 8-oxo-7,8-dihydro-2'-deoxyguanosine, and 1,N6-etheno-2'-deoxyadenosine in DNA using on-line sample preparation by HPLC column switching coupled to ESI-MS/MS.
O(6)-Methyl-2'-deoxyguanosine (O(6)-mdGuo), 8-oxo-7,8-dihydro-2'-deoxyguanosine (8-oxodGuo), and 1,N(6)-etheno-2'-deoxyadenosine (epsilondAdo) are promutagenic DNA lesions originating from both endogenous and exogenous agents and actions (methylation, hydroxylation, lipid peroxidation products). A highly sensitive quantitative method was developed to measure these DNA adducts simultaneously, using liquid chromatography tandem mass spectrometry with column switching. Deuterated O(6)-[(2)H(3)]mdGuo was synthesized and used as internal standard. The limits of quantification for O(6)-mdGuo, 8-oxodGuo, and epsilondAdo were 24, 98, and 48 fmol on column, respectively. The method showed linearity in the range 0.24-125 pmol/ml, 0.98-125 pmol/ml, and 0.49-62.5 pmol/ml for the three adducts, respectively. The inter-day precision in the linear concentration range was between 1.7 and 9.3% for O(6)-mdGuo, 10.6 and 28.7% for 8-oxodGuo, and 6.2 and 10.4%, for epsilondAdo. In DNA isolated from liver of untreated 12-week-old female F344 rats, O(6)-mdGuo was above the limit of detection (37 adducts per 10(9) normal nucleosides) but could not be quantified. 8-oxodGuo and epsilondAdo showed background levels of 500 and 130 adducts per 10(9) normal nucleosides, respectively. DNA analyzed 1h after treatment of rats with dimethylnitrosamine by oral gavage of 50 microg/kg b.wt. did not affect the levels of 8-oxodGuo and epsilondAdo but resulted in 200 O(6)-mdGuo adducts per 10(9) normal nucleosides. The method developed will be of use to study the biological significance of exogenous DNA adducts as an increment to background DNA damage and the role of modulating factors, such as DNA repair.