Merck
  • Implication of prostaglandin E(2) in TNF-alpha-induced release of m-calpain from HCS-2/8 chondrocytes. Inhibition of m-calpain release by NSAIDs.
Implication of prostaglandin E(2) in TNF-alpha-induced release of m-calpain from HCS-2/8 chondrocytes. Inhibition of m-calpain release by NSAIDs.
Osteoarthritis and cartilage (2004-10-27)
K Fushimi, S Nakashima, Y Banno, A Akaike, M Takigawa, K Shimizu
ABSTRACT
Calpains are known as Ca(2+)-dependent intracellular neutral cysteine proteases. However, m-calpain is detected in synovial fluid of arthritic joints and is shown to possess the proteoglycanase activity in vitro. The mechanism of m-calpain release into the extracellular spaces during arthritis has not yet been well characterized. In the present study, we have analyzed m-calpain release from cultured chondrocytes stimulated by a proinflammatory cytokine, tumor necrosis factor-alpha (TNF-alpha). The effects of non-steroidal anti-inflammatory drugs (NSAIDs) on m-calpain release were also examined. Human chondrocytic HCS-2/8 cells were stimulated by TNF-alpha in the presence or absence of an NSAID. m-Calpain in the cells and culture medium was quantified by Western blot analysis using an anti-m-calpain antibody. Western blots were subjected to densitometric analysis and band intensities were determined. TNF-alpha (10 ng/ml) stimulated m-calpain release with transient increase in cellular m-calpain in HCS-2/8 cells. NSAIDs examined (aspirin, loxoprofen-SRS, diclofenac sodium, indomethacin and NS398) inhibited m-calpain release and production of prostaglandin E(2) (PGE(2)) induced by 10 ng/ml TNF-alpha. Exogenously added PGE(2) accelerated the release of m-calpain in response to a lower concentration of TNF-alpha (1 ng/ml). AH6809, an EP1/2 antagonist, but not SC19220 (an EP1 antagonist), effectively inhibited TNF-alpha-induced m-calpain release. In contrast, butaprost, an EP2 agonist, accelerated release of m-calpain by 1 ng/ml TNF-alpha. These results suggest that TNF-alpha stimulates upregulation and release of m-calpain in chondrocytic HCS-2/8 cells, and that stimulation of EP2-PGE(2) receptor by produced PGE(2) is deeply involved in this process.
Sigma-Aldrich
SC 19220, ≥98% (HPLC), solid