Merck
  • N-alkyl-3-decarboxy-3-hydroxymethylsiastatin B, a new family of glycosidase inhibitors of gem-diamine 1-N-iminosugars.
N-alkyl-3-decarboxy-3-hydroxymethylsiastatin B, a new family of glycosidase inhibitors of gem-diamine 1-N-iminosugars.
Natural product letters (2002-02-13)
K I Kondo, H Adachi, Y Nishimura, T Takeuchi
ABSTRACT
N-Alkyl-3-decarboxy-3-hydroxymethylsiastatin B, N-alkyl analogues of gem-diamine 1-N-iminosugars, is a new family of glycosidase inhibitors that have been synthesized from siastatin B isolated from Streptomyces culture. These compounds were evaluated as glycosidase inhibitors.
Sigma-Aldrich
Siastatin B, lyophilized powder, from microbial