Merck
  • Diphenylpyraline-responsive parkinsonism in cerebrotendinous xanthomatosis: long-term follow up of three patients.
Diphenylpyraline-responsive parkinsonism in cerebrotendinous xanthomatosis: long-term follow up of three patients.
Journal of the neurological sciences (2001-01-04)
T Ohno, S Kobayashi, M Hayashi, M Sakurai, I Kanazawa
ABSTRACT
A long-term follow-up study was made of three patients with cerebrotendinous xanthomatosis (CTX) associated with parkinsonism, two of whom were siblings. Besides typical CTX symptoms, all three patients showed severe parkinsonism. This observation has been rarely reported in CTX. The fact that the two siblings showed parkinsonism strongly suggests the genetic propensity to parkinsonism in these CTX patients. Positron emission tomography studies of the two patients revealed presynaptic dysfunction of the nigro-striatal dopaminergic system. Treatment with the reductase inhibitor hydroxymethyl glutaryl coenzyme successfully corrected the serum cholestanol level in the early stage of the disease, which, however, did not arrest the progression of clinical symptoms, particularly their parkinsonism. Clinically, levodopa had a little effect on parkinsonism, whereas an antihistamine drug, diphenylpyraline hydrochloride (DPP) had excellent effects on all three patients throughout the long-term follow up. The mechanism of the action of DPP on parkinsonism is unclear, however, the drug seems to be a therapeutic choice for treating parkinsonism in CTX.
Sigma-Aldrich
4-Diphenylmethoxy-1-methylpiperidine hydrochloride, 95%