Merck
  • Effects of cycloheximide and aminophylline on 5-methoxytryptophol and melatonin contents in the chick pineal gland.
Effects of cycloheximide and aminophylline on 5-methoxytryptophol and melatonin contents in the chick pineal gland.
General and comparative endocrinology (2000-11-18)
J B Zawilska, J Rosiak, B Vivien-Roels, D J Skene, P Pévet, J Z Nowak
ABSTRACT
The chick pineal gland rhythmically synthesizes two 5-methoxyindoles, melatonin and 5-methoxytryptophol. These rhythms are circadian in nature and have opposite phases. The aim of this study was to determine the effects of cycloheximide, a protein synthesis inhibitor, and aminophylline, an inhibitor of phosphodiesterase, on 5-methoxytryptophol content in the chick pineal gland and to compare this with the drugs' action on pineal melatonin production. Inhibition of melatonin biosynthesis by cycloheximide (1 mg/kg, i.p. ), revealed by a marked reduction in the nighttime activity of serotonin N-acetyltransferase (AA-NAT; a key regulatory enzyme in melatonin synthesis) and melatonin concentrations, was accompanied by a significant increase in 5-methoxytryptophol content. In contrast, administration of aminophylline (100 mg/kg, i.p.) to light-exposed chicks significantly increased pineal AA-NAT activity and melatonin levels and decreased 5-methoxytryptophol concentrations. It is concluded that in the chick the production of pineal 5-methoxytryptophol and melatonin is inversely correlated.