Merck
  • Determination of daminozide in apples and apple leaves by liquid chromatography--mass spectrometry.
Determination of daminozide in apples and apple leaves by liquid chromatography--mass spectrometry.
Journal of chromatography. A (1999-03-13)
H G Mol, R C van Dam, R J Vreeken, O M Steijger
ABSTRACT
A straightforward and efficient method was developed for the determination of intact daminozide in apples and apple leaves. After extraction with methanol and a clean-up step using a graphitized carbon cartridge, the extract was analysed by ion-trap liquid chromatography-tandem mass spectrometry (LC-MS-MS) using atmospheric pressure chemical ionisation in the positive ion mode. Recoveries for apple were 98-102% with a R.S.D. < or = 11% (n = 6) and for leaves were 112-116% with a R.S.D. < or = 18% (n = 6). The limits of detection were 0.008 and 0.02 mg/kg for apples and leaves, respectively.
Product Description
Supelco
Daminozide, PESTANAL®, analytical standard