Merck
  • Incorporation of a viral DNA-packaging motor channel in lipid bilayers for real-time, single-molecule sensing of chemicals and double-stranded DNA.
Incorporation of a viral DNA-packaging motor channel in lipid bilayers for real-time, single-molecule sensing of chemicals and double-stranded DNA.
Nature protocols (2013-01-26)
Farzin Haque, Jia Geng, Carlo Montemagno, Peixuan Guo
ABSTRACT
Over the past decade, nanopores have rapidly emerged as stochastic biosensors. This protocol describes the cloning, expression and purification of the channel of the bacteriophage phi29 DNA-packaging nanomotor and its subsequent incorporation into lipid membranes for single-pore sensing of double-stranded DNA (dsDNA) and chemicals. The membrane-embedded phi29 nanochannel remains functional and structurally intact under a range of conditions. When ions and macromolecules translocate through this nanochannel, reliable fingerprint changes in conductance are observed. Compared with other well-studied biological pores, the phi29 nanochannel has a larger cross-sectional area, which enables the translocation of dsDNA. Furthermore, specific amino acids can be introduced by site-directed mutagenesis within the large cavity of the channel to conjugate receptors that are able to bind specific ligands or analytes for desired applications. The lipid membrane-embedded nanochannel system has immense potential nanotechnological and biomedical applications in bioreactors, environmental sensing, drug monitoring, controlled drug delivery, early disease diagnosis and high-throughput DNA sequencing. The total time required for completing one round of this protocol is around 1 month.
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Xylene Cyanol FF, BioReagent, Molecular Biology
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand
Avanti
PC Membranes 0.1um, Avanti Research - A Croda Brand
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand