Merck
  • [Interest of an intensive chemotherapy for intravascular large B cell lymphoma].
[Interest of an intensive chemotherapy for intravascular large B cell lymphoma].
Bulletin du cancer (2013-04-17)
Aurélie Baldolli, Marie Chuffart, Loik Geffray, Laurence Verneuil, Oumédaly Reman
ABSTRACT
We describe three cases of intravascular lymphoma B with different clinical presentation: one case of a cutaneous variant and two cases with surrenal and cutaneous localisation. All patients are in complete remission after chemotherapy alone or after chemotherapy and autologous stem cells transplantation. The review of the literature as well as our cases specify the interest of an aggressive chemotherapy with autologous of peripheral stem cells if it was possible.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, suitable for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Vindesine sulfate salt hydrate, ≥95% (HPLC)