Merck
  • Optimization of ABTS radical cation assay specifically for determination of antioxidant capacity of intracellular extracts of microalgae and cyanobacteria.
Optimization of ABTS radical cation assay specifically for determination of antioxidant capacity of intracellular extracts of microalgae and cyanobacteria.
Food chemistry (2012-12-26)
A Catarina Guedes, Helena M Amaro, Maria S Gião, F Xavier Malcata
ABSTRACT
A renewed interest in antioxidants has arisen in recent years; microalgae and cyanobacteria are potential sources thereof for use as food/feed ingredients. However, improved methods for comprehensive screening of antioxidant capacity specifically in intracellular extracts of marine microorganisms are required - encompassing lipophilic and hydrophilic compounds simultaneously. The original ABTS method was thus improved, and in particular the procedures of cell disruption and storage were optimized. The best solvent found was ethanol/water (1:1, v/v). The reaction to form ABTS(+) in said solvent was essentially complete by eight hours, and this radical cation was stable for at least 6 days; at room temperature, the ABTS(+) solution remained within an allowable analytical range for up to 13 h. Ultra Turrax was the best cell disruption method, and refrigeration was the best preservation method. This improved methodology was validated with four representative strains that respond poorly to cell disruption.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Pricing and availability is not currently available.