Merck
  • The naturally occurring aliphatic isothiocyanates sulforaphane and erucin are weak agonists but potent non-competitive antagonists of the aryl hydrocarbon receptor.
The naturally occurring aliphatic isothiocyanates sulforaphane and erucin are weak agonists but potent non-competitive antagonists of the aryl hydrocarbon receptor.
Archives of toxicology (2012-05-31)
Ahmad F Abdull Razis, Natalya Hanlon, Ewa Soltys, Veronika Krizova, Renato Iori, Kathryn E Plant, Nick Plant, Costas Ioannides
ABSTRACT
As the Ah receptor target gene products play a critical role in chemical carcinogenesis, antagonists are considered as potential chemopreventive agents. It is demonstrated in this paper that the isothiocyanates R,S-sulforaphane and erucin are non-competitive antagonists of the aryl hydrocarbon (Ah) receptor. Both isothiocyanates were poor agonists for the receptor and elevated CYP1A1 mRNA levels only modestly when incubated with precision-cut rat liver slices. In contrast, the classical Ah receptor agonist benzo[a]pyrene was a potent inducer of CYP1A1 mRNA levels, with this effect being effectively antagonized by the two isothiocyanates. In further studies, it was demonstrated that R,S-sulforaphane could both prevent the interaction of and displace already bound benzo[a]pyrene from the Ah receptor, but no concentration dependency was observed with respect to the isothiocyanate. Both erucin and R,S-sulforaphane antagonized the benzo[a]pyrene-mediated increase in the CYP1A-mediated O-deethylation of ethoxyresorufin in rat precision-cut liver slices. Of the two isomers of R,S-sulforaphane, the naturally occurring R-isomer was more effective than the S-isomer in antagonizing the activation of the Ah receptor by benzo[a]pyrene. Antagonism of the Ah receptor may be a major contributor to the established chemoprevention of aliphatic isothiocyanates.
Sigma-Aldrich
Resorufin ethyl ether, suitable for fluorescence, ≥95% (UV)
Sigma-Aldrich
Resorufin ethyl ether, ≥98% (TLC), powder