Merck
  • Further structure-activity relationship studies on 8-substituted-3-[2-(diarylmethoxyethylidenyl)]-8-azabicyclo[3.2.1]octane derivatives at monoamine transporters.
Further structure-activity relationship studies on 8-substituted-3-[2-(diarylmethoxyethylidenyl)]-8-azabicyclo[3.2.1]octane derivatives at monoamine transporters.
Bioorganic & medicinal chemistry (2011-11-08)
Shaine A Cararas, Sari Izenwasser, Dean Wade, Amy Housman, Abha Verma, Stacey A Lomenzo, Mark L Trudell
ABSTRACT
The synthesis and structure-activity relationships of 8-substituted-3-[2-(diarylmethoxyethylidenyl)]-8-azabicyclo[3.2.1]octane derivatives were investigated at the dopamine transporter (DAT), the serotonin transporter (SERT) and norepinephrine transporter (NET). The rigid ethylidenyl-8-azabicyclic[3.2.1]octane skeleton imparted modestly stereoselective binding and uptake inhibition at the DAT. Additional structure-activity studies provided a transporter affinity profile that was reminiscent of the structure-activity of GBR 12909. From these studies, the 8-cyclopropylmethyl group has been identified as a unique moiety that imparts high SERT/DAT selectivity. In this study the 8-cyclopropylmethyl derivative 22e (DAT K(i) of 4.0 nM) was among the most potent compounds of the series at the DAT and was the most DAT selective ligand of the series (SERT/DAT: 1060). Similarly, the 8-chlorobenzyl derivative 22g (DAT K(i) of 3.9 nM) was found to be highly selective for the DAT over the NET (NET/DAT: 1358).
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Octane, anhydrous, ≥99%
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, reagent grade, 98%