Merck
  • Can microorganisms convert antimony trioxide or potassium antimonyl tartrate to methylated stibines?
Can microorganisms convert antimony trioxide or potassium antimonyl tartrate to methylated stibines?
The Science of the total environment (1997-12-31)
P N Gates, H A Harrop, J B Pridham, B Smethurst
ABSTRACT
No evidence could be found for the production, in culture, of methylated antimony compounds from water-insoluble or soluble antimony derivatives by the aerobes, Scopulariopsis brevicaulis or Bacillus sp. or by anaerobes associated with cot mattress materials. The study does not support the hypothesis that volatile organoantimony compounds are a cause of cot deaths. Anaerobic cultures from a polluted pond generated trimethylstibine from potassium antimonyl tartrate.
Sigma-Aldrich
Antimony(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, nanopowder, <250 nm particle size (TEM), ≥99.9% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, powder, 5 μm, ReagentPlus®, 99%