Merck
  • Use of a triple-stain technique to detect viability and acrosome reaction in deer spermatozoa.
Use of a triple-stain technique to detect viability and acrosome reaction in deer spermatozoa.
Archives of andrology (1997-07-01)
J J Garde, N Ortiz, A García, L Gallego
ABSTRACT
A procedure is described for simultaneously evaluating of sperm viability and acrosomal status in fixed deer spermatozoa. This technique, which is a modification of the triple-stain technique (TST) developed for human sperm, utilizes trypan blue to identify the percentage of live sperm in culture and subsequent staining of fixed sperm to differentiate the acrosome. Four classes of deer sperm can be distinguished with the TST: (1) live unreacted sperm, (2) live acrosome-reacted sperm (true acrosome reaction), (3) dead unreacted sperm, and (4) dead acrosome-reacted sperm (false acrosome reaction). The study shows that the acrosome status can be detected either by TST stain or by phase-contrast microscopy (r = .971, p < .001) and that TST does not differ from 0.4% trypan blue in its ability to identify live and dead sperm (r = .995, p < .001). This staining procedure enables differentiation of spermatozoa that have undergone a true acrosome reaction (TAR) from those that have undergone a false acrosome reaction (FAR). In this way, incubation of deer spermatozoa for 30 min at 37 degrees C in the presence of calcium ionophore A23187 increased the proportion of spermatozoa undergoing a TAR. Sperm incubated in the absence of A23187 undergo the TAR at a much lower rate (p < .001) than those incubated in the presence of ionophore. This stain procedure was also used to study the time course of the true acrosome reaction of deer sperm in vitro. Incubation of deer spermatozoa for up 24 h at 37 degrees C resulted in a decrease in the percentage of live acrosome-intact spermatozoa and a simultaneous increase in the percentage of spermatozoa categorized as having undergone a TAR and FAR.
Sigma-Aldrich
Bismarck Brown Y, certified by the BSC, Dye content 50 %