Merck
  • Simultaneous determination of the colorants sunset yellow FCF and quinoline yellow by solid-phase spectrophotometry using partial least squares multivariate calibration.
Simultaneous determination of the colorants sunset yellow FCF and quinoline yellow by solid-phase spectrophotometry using partial least squares multivariate calibration.
The Analyst (1997-04-01)
L F Capitán-Vallvey, M D Fernández, I de Orbe, J L Vilchez, R Avidad
ABSTRACT
A method for the simultaneous determination of the colorants Sunset Yellow FCF and Quinoline Yellow using solid-phase spectrophotometry is proposed. The colorants were isolated in Sephadex DEAE A-25 gel at pH 5.0, the gel-colorants system was packed in a 1 mm silica cell and spectra were recorded between 400 and 600 nm against a blank. Statistical results were obtained by partial least squares (PLS) multivariate calibration. The optimized matrix by using the PLS-2 method enables the determination of the colorants in artificial mixtures and commercial soft drinks.
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)
Sigma-Aldrich
Quinoline Yellow, Mixture of the mono- and disulfonic acids of Quinoline Yellow
Sigma-Aldrich
Quinoline Yellow, mixture of mono- and disulfonic acid sodium salt