Merck
  • Substitution effect of purine nucleosides on diisobutylaluminum hybride reduction.
Substitution effect of purine nucleosides on diisobutylaluminum hybride reduction.
Nucleic acids symposium series (1995-01-01)
K Hirota, Y Monguchi, M Sako, Y Kitade
ABSTRACT
It is reported that the diisobutylaluminum hydride (DIBALH) reduction of inosine and adenosine derivatives (1a and 1d) causes cleavage of the ribose moiety to give the corresponding 9-ribitylhypoxanthine (2a) and 9-ribityladenine (2d), respectively. The substitution effect of purine nucleosides on the reduction was investigated. O6-alkylinosine derivatives (1b and 1c) were reduced in good yields like 1a. Introduction of methyl group into the N6-position of adenosine (1d), however, greatly reduced the reactivity toward DIBALH. The reduction of 7-deazainosine (3a) and 7-deazaadenosine (3b) hardly proceeded. These results suggest that oxygenophilicity of aluminum atom facilitates coordination of DIBALH to the 7-ring nitrogen atom and accelerate a cleavage of the ribose.
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in heptane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in hexanes
Sigma-Aldrich
Diisobutylaluminum hydride, reagent grade
Sigma-Aldrich
Diisobutylaluminum hydride solution, 25 wt. % in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in cyclohexane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in toluene