Merck
  • Imaging of DNA in human and plant chromosomes by high-resolution scanning electron microscopy.
Imaging of DNA in human and plant chromosomes by high-resolution scanning electron microscopy.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology (1995-09-01)
G Wanner, H Formanek
ABSTRACT
We describe a DNA-specific staining procedure, using a blue platinum organic dye, which allows DNA imaging of chromosomes by detection of back-scattered electrons in the scanning electron microscope. DNA distribution is visualized within chromosomal details of the centromeric and satellite regions, or in interphase chromatin, with high-resolution (10-15 nm) for comparison with the corresponding secondary electron image representing DNA plus protein.
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 98%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), ≥99.9% trace metals basis
Sigma-Aldrich
Ammonium tetrachloroplatinate(II), 99%