Merck
  • [Comparative studies of 35S-hemineurine and 35S-thiamine distribution and binding by proteins in the neural structures of the rat brain].
[Comparative studies of 35S-hemineurine and 35S-thiamine distribution and binding by proteins in the neural structures of the rat brain].
Voprosy meditsinskoi khimii (1980-09-01)
L B Filippova, A Ia Rozanov
ABSTRACT
35S-Hemineurine penetrated across the blood-brain barrier more rapidly than 35S-thiamin within the first 30 min after intravenous administration. Within 24 hrs content of 35S-thiamin in the neurones exceeded by 34% its concentration in glia and by 200% -- in myelin fraction. 35S-Hemineurine was bound slightly with brain proteins as compared with thiamin and it was removed at the higher rate from brain regions and subcellular structures.
Sigma-Aldrich
4-Methyl-5-thiazoleethanol, 98%
Sigma-Aldrich
4-Methyl-5-thiazoleethanol, ≥98%, FG
Supelco
4-Methyl-5-thiazoleethanol, analytical standard