Merck
  • Effects of phorbol ester tumor promoters and hyperplasiogenic agents on cytoplasmic glucocorticoid receptors in epidermis.
Effects of phorbol ester tumor promoters and hyperplasiogenic agents on cytoplasmic glucocorticoid receptors in epidermis.
The Journal of investigative dermatology (1982-12-01)
K A Davidson, T J Slaga
ABSTRACT
Glucocorticoids (anti-inflammatory steroids) are very potent inhibitors of mouse skin tumor promotion induced by 12-O-tetradecanoylphorbol-13-acetate (TPA). This report describes a high-affinity, limited-capacity binding component which specifically interacts with glucocorticoids and which is identified as a glucocorticoid (GC) receptor present in the cytosol of adult mouse epidermis. Also described is the effect of a single application of TPA and other hyperplasiogenic agents on the level of epidermal cytosol GC receptor. After treatment with acetone the epidermal GC receptor level was 0.38 pmol/mg cytosol protein, whereas after application of TPA (4 micrograms) there was a time-dependent transient decrease in the level of GC receptor (39% of control 24 hr after TPA) followed by an increase (55% and 66% of control 48 and 72 hr, respectively, after TPA). Hyperplasiogenic agents such as mezerein and ethylphenylpropiolate were also effective in causing a reduction in the level of epidermal cytosol GC receptor level, but weak tumor promoters such as phorbol dibenzoate and 4-O-methyl TPA were less effective than TPA. Therefore, there is a good correlation among agents which induce epidermal hyperplasia and agents which cause a reduction in epidermal cytosol GC receptor levels. The functional significance of the decrease in receptor level is discussed.
Sigma-Aldrich
Ethyl phenylpropiolate, 98%