Merck
  • Effects on plasma glucose concentration of light-for-date infants and infants of diabetic mothers of feeds supplemented with a glucose polymer.
Effects on plasma glucose concentration of light-for-date infants and infants of diabetic mothers of feeds supplemented with a glucose polymer.
Acta paediatrica Scandinavica (1985-05-01)
C Costalos, G Russell, Q Al Rahim, M J Tarlow, D J Lloyd, K R Ross
ABSTRACT
Serial plasma glucose estimations were performed in 44 light-for-date infants and 17 infants of diabetic mothers fed 2, 3 or 4-hourly with feeds containing 10% additional carbohydrate in the form of a glucose polymer (Caloreen). In the infants fed 2 and 3-hourly, plasma glucose was higher following the high carbohydrate feeds, particularly immediately before the next feed was due. Given 4-hourly, the high carbohydrate feed had little effect in most light-for-date infants, and in one light-for-date infant and two infants of diabetic mothers the plasma glucose was lower at the end of 4 hours than following normal formula. It is concluded that the addition of glucose polymer to feeds given 2 and 3 hourly has a potentially useful effect in neonates at risk of developing hypoglycaemia, but it cannot be used to increase the feed interval to 4 hours.
Product Description
Millipore
Dextrin from potato starch, suitable for microbiology
Sigma-Aldrich
Dextrin from corn, commercial grade, Type II, powder
Millipore
Dextrin from potato starch, for biotechnological purposes
Sigma-Aldrich
Dextrin from corn, Type I, powder