Merck
An improved fluorimetric assay for brain monoamine oxidase.
Journal of pharmacological methods (1985-06-01)
A Morinan, H M Garratt
ABSTRACT
A fluorimetric assay for monamine oxidase that uses kynuramine as the substrate is described. This method is more economic, rapid, reproducible, and sensitive than other similar procedures. Its validity has been confirmed by the study of the subcellular distribution, the effects of inhibitors, and the kinetics of enzyme activity. Previous reports of substrate inhibition by kynuramine may have to be reassessed in the light of evidence presented here.
Sigma-Aldrich
4-Quinolinol, 98%