Merck
  • Effect of cortisol on neurophysin I/oxytocin and peptidyl glycine-alpha-amidating mono-oxygenase mRNA expression in bovine luteal and granulosa cells.
Effect of cortisol on neurophysin I/oxytocin and peptidyl glycine-alpha-amidating mono-oxygenase mRNA expression in bovine luteal and granulosa cells.
Polish journal of veterinary sciences (2013-08-27)
A Ziolkowska, J Mlynarczuk, J Kotwica
ABSTRACT
Cortisol stimulates the synthesis and secretion of oxytocin (OT) from bovine granulosa and luteal cells, but the molecular mechanisms of cortisol action remain unknown. In this study, granulosa cells or luteal cells from days 1-5 and 11-15 of the oestrous cycle were incubated for 4 or 8 h with cortisol (1 x 10(-5), 1 x 10(-7) M). After testing cell viability and hormone secretion (OT, progesterone, estradiol), we studied the effect of cortisol on mRNA expression for precursor of OT (NP-I/OT) and peptidyl glycine-alpha-amidating mono-oxygenase (PGA). The influence of RU 486 (1 x 10(-5) M), a progesterone receptor blocker and inhibitor of the glucocorticosteroid receptor (GR), on the expression for both genes was tested. Cortisol increased the mRNA expression for NP-I/OT and PGA in granulosa cells and stimulated the expression for NP-I/OT mRNA in luteal cells obtained from days 1-5 and days 11-15 of the oestrous cycle. Expression for PGA mRNA was increased only in luteal cells from days 11-15 of the oestrous cycle. In addition, RU 486 blocked the cortisol-stimulated mRNA expression for NP-I/OT and PGA in both types of cells. These data suggest that cortisol affects OT synthesis and secretion in bovine ovarian cells, by acting on the expression of key genes, that may impair ovary
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Oxytocin, European Pharmacopoeia (EP) Reference Standard