Merck
  • Preparation of nanoscale Ni-B amorphous alloys and their application in the selective hydrogenation of cinnamic acid.
Preparation of nanoscale Ni-B amorphous alloys and their application in the selective hydrogenation of cinnamic acid.
Journal of nanoscience and nanotechnology (2013-08-02)
Guoyi Bai, Huixian Dong, Zhen Zhao, Yalong Wang, Qingzhi Chen, Mande Qiu
ABSTRACT
A series of metal modified nanoscale Ni-B amorphous alloys was prepared by chemical reduction and tested in the selective hydrogenation of cinnamic acid. A Co modified Ni-B amorphous alloy (Ni-Co-B) exhibited excellent catalytic performance in this reaction with both 100.0% conversion of cinnamic acid and 100.0% selectivity for hydrocinnamic acid under the optimized reaction conditions. X-ray diffraction (XRD) results indicated that the addition of Co had not changed the amorphous structure of Ni-B; whereas, its addition was believed not only to favor decreased agglomeration of the active Ni species, as proven by transmission electron microscopy (TEM), but also to contribute to adsorption of hydrogen itself. Thus, Ni-Co-B showed a larger BET surface area, smaller particle size, and greater number of active species resulting in optimum H2-chemisorption compared to Ni-B and accounting for its excellent catalytic performance in cinnamic acid hydrogenation.
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Supelco
trans-Cinnamic acid, analytical standard
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
Hydrogen, ≥99.99%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis