Merck
  • ZnO piezoelectric fine wire gated graphene oxide field effect transistor.
ZnO piezoelectric fine wire gated graphene oxide field effect transistor.
Journal of nanoscience and nanotechnology (2013-07-19)
Rajneesh Mohan, Karthikeyan Krishnamoorthy, Gui-Sik Kim, Sang-Jae Kim
ABSTRACT
Here we report the fabrication and characteristics of graphene oxide (GO) field effect transistor gated with piezopotential of ZnO fine wires on a flexible substrate. The FET device consists of GO thin film on the bottom and ZnO piezoelectric fine wire (PFW) on the top. In the FET device the GO serves as a carrier transport channel and ZnO PFW acts as a gate. When the substrate is bent, a piezopotential is generated in the ZnO PFW. The piezopotential created by the strain in the ZnO PFW was used to control the carrier transport in the GO channel. This device demonstrates the application of piezoelectric ZnO PFW for creating the gating effect on the semiconducting performance of GO film.
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%