Merck
  • Remineralisation of occlusal white spot lesion with a combination of 10% CPP-ACP and 0.2% sodium fluoride evaluated using Diagnodent: a pilot study.
Remineralisation of occlusal white spot lesion with a combination of 10% CPP-ACP and 0.2% sodium fluoride evaluated using Diagnodent: a pilot study.
Oral health & preventive dentistry (2013-06-13)
Jogikalmat Krithikadatta, Chain Fredrick, Mohan Abarajithan, Deivanayagam Kandaswamy
ABSTRACT
To evaluate the efficacy of 10% casein phosphopeptide amorphous calcium phosphate complex (CPP-AC P) used alone or with fluoride as compared to fluoride mouthrinse for the remineralisation of occlusal white spot lesions in an in vivo pilot study. Forty-five subjects with occlusal white spot lesions (WSL) determined by visual examination and confirmed with Diagnodent were randomly divided into 3 groups: group 1: 10% CPP-AC P + 0.2% NaF; group 2: 10% CPPAC P; group 3: 0.5% NaF mouthrinse for 30 days. The pre-treatment and post-intervention Diagnodent values were tabulated and subjected to statistical analysis. Among the groups, the remineralising potential of 10% CPP-AC P + 0.2% NaF (group 1) and 10% CPP-AC P (group 2) were similar, however, both groups were superior to 0.5% NaF mouthrinse (group 3). The CCP-AC P technique is highly beneficial for the remineralisation of non-cavitated occlusal white spot lesions when compared to 0.5% NaF mouthrinse.
Sigma-Aldrich
Sodium Fluoride Solution
Supelco
Fluoride ion solution for ISE, for ion-selective electrodes, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%