Merck
  • Biodegradation of benzo[a]pyrene by the mixed culture of Bacillus cereus and Bacillus vireti isolated from the petrochemical industry.
Biodegradation of benzo[a]pyrene by the mixed culture of Bacillus cereus and Bacillus vireti isolated from the petrochemical industry.
Journal of environmental biology (2013-06-08)
Ramya Mohandass, Pallabi Rout, Sonia Jiwal, Chitrambalam Sasikala
ABSTRACT
Polycyclic aromatic hydrocarbons are a group of compounds that pose threat to humans and animal life. Methods to reduce the amount of PAHs in the environment are continuously being sought. The bacterial consortium capable of utilizing benzo(a)pyrene as the sole source of carbon and energy was isolated from petrochemical soil. The isolates were identified as Bacillus cereus and Bacillus viretibased on morphological characterization, and 16S rDNA gene sequence analysis. About 58.98% of benzo(a)pyrene at concentration of 500 mg l(-1) in mineral salts medium were removed by bacterial consortium. GC mass spectral analysis showed the presence of metabolite cis-4-(7-hydroxypyren-8-yl)-2-oxobut-3enoic acid. The results indicate that the bacterial consortium is a new bacterial resource for biodegrading benzo(a)pyrene and might be used for bioremediation of sites heavily contaminated by benzo[a]pyrene and its derivatives.
Product Description
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis