Merck
  • Short-course prophylactic zinc supplementation for diarrhea morbidity in infants of 6 to 11 months.
Short-course prophylactic zinc supplementation for diarrhea morbidity in infants of 6 to 11 months.
Pediatrics (2013-06-05)
Akash Malik, Davendra K Taneja, Niveditha Devasenapathy, K Rajeshwari
ABSTRACT
Zinc supplementation during diarrhea substantially reduces the incidence and severity of diarrhea. However, the effect of short-course zinc prophylaxis has been observed only in children >12 months of age. Because the incidence of diarrhea is comparatively high in children aged 6 to 11 months, we assessed the prophylactic effect of zinc on incidence and duration of diarrhea in this age group. In this randomized, double-blind, placebo-controlled trial, we enrolled infants aged 6 to 11 months from an urban resettlement colony in Delhi, India, between January 1, 2011, and January 15, 2012. We randomly assigned 272 infants to receive either 20 mg of zinc or a placebo suspension orally every day for 2 weeks. The primary outcome was the incidence of diarrhea per child-year. All analyses were done by intention-to-treat. A total of 134 infants in the zinc and 124 in the placebo groups were assessed for the incidence of diarrhea. There was a 39% reduction (crude incident rate ratio [IRR] 0.61, 95% confidence interval [CI] 0.53-0.71) in episodes of diarrhea, 39% (adjusted IRR 0.61, 95% CI 0.54-0.69) in the total number of days that a child suffered from diarrhea, and reduction of 36% in duration per episode of diarrhea (IRR 0.64, 95% CI 0.56-0.74) during the 5 months of follow-up. Short-course prophylactic zinc supplementation for 2 weeks may reduce diarrhea morbidity in infants of 6 to 11 months for up to 5 months, in populations with high prevalence of wasting and stunting.
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate solution, 0.3 N