Merck
  • [Evaluation on preparation process of brevisapin colon-specific tables and its in vitro release].
[Evaluation on preparation process of brevisapin colon-specific tables and its in vitro release].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-31)
Yan-Ling Li, Jin-Yu Wang, De-Bin Yang, Mei-Hong Dong, Yan Tong
ABSTRACT
To prepare the new traditional Chinese medicine preparation--pH-dependent brevisapin colon-specific tablets, and investigate its in vitro release, in order to discuss the feasibility of preparing colon-targeted traditional Chinese medicines. With scutellarin, the active ingredient in brevisapin, as the evaluation index, coating prescriptions of the preparation was screened. The in vitro release determination method was adopted to detect the in vitro release performance of the preparation. The in vitro release determination results showed no brevisapin in artificial pH 1. 2 dilute hydrochloric acid solution for 2 h, an accumulated dissolution rate of less than 5% in pH 6. 8 phosphate buffer solution for 4 h, but an accumulated dissolution rate exceeding 90% in pH 7. 6 phosphate buffer solution for 1 h. Brevisapin colon-specific tablets prepared can realize colon-specific release.
Product Description
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Apigenin, analytical standard
Supelco
Scutellarin, analytical standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur