Merck
  • Relationship between occupational exposure to cadmium, transaminases and γ-GT in workers exposed to urban stressors.
Relationship between occupational exposure to cadmium, transaminases and γ-GT in workers exposed to urban stressors.
Annali di igiene : medicina preventiva e di comunita (2013-05-25)
F Tomei, M Ciarrocca, M V Rosati, T Casale, C Di Pastena, H A Nieto, V Antuono, G Iannattone, G Tomei, T Caciari
ABSTRACT
The exposure to Cd has been linked to many disease and liver is the major target organ of Cd toxicity following acute or chronic exposure.No study has ever evaluated the association between liver function tests and exposure to Cd present in urban air in outdoor workers. In order to assess the relationship between the exposure to urban Cd and the associated changes in liver functionality markers, we conducted a study on 110 municipal policemen divided into 2 groups according to their tasks: 60 traffic policemen and 50 drivers, underwent tests for the determination of the urinary values of Cd correlated with the blood levels of transaminases and ?-GT. Moreover we investigated the exposure to Cd in the air through a personal dosimetry on a subgroup of 40 subjects (20 traffic policemen 20 police drivers). The results showed that the average personal exposure to air Cd was significantly higher in traffic policemen than drivers. Furthermore, in the subset of 40 subjects and in the total sample of 110 subjects, the mean values of urinary Cd were significantly higher in traffic policemen rather than in the police drivers. No significant difference was observed in the mean values of liver functionality in both groups. Due to the few studies on the effects of the exposure to Cd in big cities, the results of the present study offers useful information about occupational exposure to Cd on the road and in the car and the lack of effects on liver function.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid