Merck
  • Prevalence of the lmo0036-0043 gene cluster encoding arginine deiminase and agmatine deiminase systems in Listeria monocytogenes.
Prevalence of the lmo0036-0043 gene cluster encoding arginine deiminase and agmatine deiminase systems in Listeria monocytogenes.
The new microbiologica (2013-05-21)
Jianshun Chen, Fan Chen, Changyong Cheng, Weihuan Fang
ABSTRACT
Arginine deiminase and agmatine deiminase systems are involved in acid tolerance, and their encoding genes form the cluster lmo0036-0043 in Listeria monocytogenes. While lmo0042 and lmo0043 were conserved in all L. monocytogenes strains, the lmo0036-0041 region of this cluster was identified in all lineages I and II, and the majority of lineage IV (83.3%) strains, but absent in all lineage III and a small fraction of lineage IV (16.7%) strains, suggesting that the presence of the complete lmo0036-0043 cluster is dependent on lineages. lmo0036-0043-complete and -deficient lineage IV strains exhibit specific ascB-dapE profiles, which might represent two subpopulations with distinct genetic characteristics.
Sigma-Aldrich
N,N′-Bis(2-aminoethyl)-1,3-propanediamine, 97%