Merck
  • The mood stabilizer valproate activates human FGF1 gene promoter through inhibiting HDAC and GSK-3 activities.
The mood stabilizer valproate activates human FGF1 gene promoter through inhibiting HDAC and GSK-3 activities.
Journal of neurochemistry (2013-05-08)
Chien-Yu Kao, Yi-Chao Hsu, Jen-Wei Liu, Don-Ching Lee, Yu-Fen Chung, Ing-Ming Chiu
ABSTRACT
Valproic acid (VPA) is the primary mood-stabilizing drug to exert neuroprotective effects and to treat bipolar disorder in clinic. Fibroblast growth factor 1 (FGF1) has been shown to regulate cell proliferation, cell division, and neurogenesis. Human FGF1 gene 1B promoter (-540 to +31)-driven green fluorescence (F1BGFP) has been shown to recapitulate endogenous FGF1 gene expression and facilitates the isolation of neural stem/progenitor cells (NSPCs) from developing and adult mouse brains. In this study, we provide several lines of evidence to demonstrate the underlying mechanisms of VPA in activating FGF-1B promoter activity: (i) VPA significantly increased the FGF-1B mRNA expression and the percentage of F1BGFP(+) cells; (ii) the increase of F1BGFP expression by VPA involves changes of regulatory factor X (RFX) 1-3 transcriptional complexes and the increase of histone H3 acetylation on the 18-bp cis-element of FGF-1B promoter; (iii) treatments of other histone deacetylases (HDAC) inhibitors, sodium butyrate and trichostatin A, significantly increased the expression levels of FGF-1B, RFX2, and RFX3 transcripts; (iv) treatments of glycogen synthase kinase 3 (GSK-3) inhibitor, lithium, or GSK-3 siRNAs also significantly activated FGF-1B promoter; (v) VPA specifically enhanced neuronal differentiation in F1BGFP(+) embryonic stem cells and NSPCs rather than GFP(-) cells. This study suggested, for the first time, that VPA activates human FGF1 gene promoter through inhibiting HDAC and GSK-3 activities.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-Galactocerebroside Antibody, clone mGalC, clone mGalC, Chemicon®, from mouse
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-phospho-GSK3β (Ser9) Antibody, clone 2D3, clone 2D3, Upstate®, from mouse
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Anti-MAP2A Antibody, AP20, ascites fluid, clone AP20, Chemicon®
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit