Merck
  • Incorporation of Ag metallic nanoparticles in 3D gelatin matrix via the green strategy solution plasma.
Incorporation of Ag metallic nanoparticles in 3D gelatin matrix via the green strategy solution plasma.
Journal of nanoscience and nanotechnology (2013-05-08)
Panuphong Pootawang, Seong Cheol Kim, Jung Wan Kim, Sang Yul Lee
ABSTRACT
The environmental concern pays much attention to the recent cause of the global warming effect. The reduction of the chemical uses is one of many ways to avoid this crucial problem. Herein, the green process for silver nanometallic particle formation and incorporation in gelatin are proposed. By using a novel discharge process in solution named solution plasma, the silver nanometallic particle formation and its incorporation in gelatin could be accomplished in one-batch reactor during discharge by using silver nitrate (AgNO3) solution as the precursor and controlling systematical parameters. The three-dimensional scaffolds of gelatin/silver biocomposite were fabricated using lyophilizer and the water-soluble property of gelatin was improved by irradiation of ultraviolet ray. The well dispersed silver nanoparticles with the mean particle size 10-20 nm in the good texture of gelatin matrix were obtained. The density of micropore in gelatin/silver scaffold was proportional to the gelatin concentration. In addition, thermal stability of prepared samples had no change comparing with pure gelatin, indicating that the incorporation of silver nanoparticles in gelatin matrix did not affect to the nature of gelatin.
Product Description
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture