Merck
[Nicotine replacement therapy].
Nihon rinsho. Japanese journal of clinical medicine (2013-05-02)
Shunnosuke Handa
ABSTRACT
For the habitual smokers with nicotine dependence, it is difficult for them to quit smoking. The nicotine replacement therapy (NRT) is a helpful measure for the persons who want to quit smoking. The concept, medication and practice of the NRT are described.
Product Description
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid