Merck
  • Computational study on the geometry optimization and excited - state properties of riboflavin by ArgusLab 4.0.1.
Computational study on the geometry optimization and excited - state properties of riboflavin by ArgusLab 4.0.1.
Pakistan journal of pharmaceutical sciences (2013-04-30)
Ambreen Hafeez, Afshan Naz, Sadaf Naeem, Khalida Bano, Naheed Akhtar
ABSTRACT
Riboflavin (vitamin B2) belongs to a group of respiratory enzymes that occur widely in animals and plants participating in vital oxidation- reduction processes in the body. A computational study was conducted on riboflavin by ArgusLab 4.0.1 to obtain the most active conformation of riboflavin and to analyze its excited-state properties. The best conformation of riboflavin was found to be -199.2173 kcal/mol which is the minimum potential energy calculated by geometry convergence function by ArgusLab software; performed according to Hartree-Fock calculation method. Electronic transition states (ground and excited), were also calculated and visualized by semi-empirical ZINDO method by ArgusLab from which molecular properties such as energies, wave function and dipole moments were established. All the results obtained from geometry optimization and excited-state properties lead us to delineate the active sites with charged groups of riboflavin to interact with the receptors. Such types of investigations are significant for drug-receptor interactions.
Product Description
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Supelco
Riboflavin (B2), analytical standard