Merck
  • Evaluation of poly(ethylene glycol)/hydroxypropyl starch aqueous two-phase system for immunoglobulin G extraction.
Evaluation of poly(ethylene glycol)/hydroxypropyl starch aqueous two-phase system for immunoglobulin G extraction.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-04-23)
Qiang Wu, Dong-Qiang Lin, Shan-Jing Yao
ABSTRACT
The aqueous two-phase system (ATPS) with polyethylene glycol (PEG) and hydroxypropyl starch (HPS) was evaluated for the extraction of human immunoglobulin G (IgG) from human serum albumin (HSA). The partitions of IgG in PEG/sulphate, PEG/phosphate, PEG/Dextran and PEG/HPS ATPSs were compared, and the results indicated that PEG/HPS system was most suitable for IgG extraction. The effects of the concentrations of PEG, HPS and NaCl addition and pH on the partition of IgG and HSA in PEG/HPS ATPS were investigated. It was found that with 15% NaCl addition at pH 8.0 IgG could be largely recovered in the top PEG-rich phase and most of HSA kept in the bottom phase. In addition, the response surface methodology (RSM) was used to optimize the extraction of IgG from the protein mixture of IgG and HSA. The optimal conditions were obtained as 12% (w/w) PEG 4000, 18% (w/w) HPS and 10% (w/w) NaCl at pH 8.0. The extraction yield of IgG in the top phase was 99.2% and the purification factor could reach 5.28. The back extractions of IgG into a phosphate-rich bottom phase were also studied. The total purification factor was 5.73 with the yield of 84.0%. The results indicated that PEG/HPS ATPS might be a promising alternative for the primary recovery of IgG from the complicated feedstock.
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble