Merck
  • Slow freezing coupled static magnetic field exposure enhances cryopreservative efficiency--a study on human erythrocytes.
Slow freezing coupled static magnetic field exposure enhances cryopreservative efficiency--a study on human erythrocytes.
PloS one (2013-03-23)
Chun-Yen Lin, Po-Li Wei, Wei-Jen Chang, Yung-Kai Huang, Sheng-Wei Feng, Che-Tong Lin, Sheng-Yang Lee, Haw-Ming Huang
ABSTRACT
The aim of this study was to assess the cryoprotective effect of static magnetic fields (SMFs) on human erythrocytes during the slow cooling procedure. Human erythrocytes suspended in 20% glycerol were slowly frozen with a 0.4-T or 0.8-T SMF and then moved to a -80°C freezer for 24 hr. The changes in survival rate, morphology, and metabolites of the thawed erythrocytes were examined. To understand possible cryoprotective mechanisms of SMF, membrane fluidity and dehydration stability of SMF-exposed erythrocytes were tested. For each test, sham-exposed erythrocytes were used as controls. Our results showed that freezing coupled with 0.4-T or 0.8-T SMFs significantly increased the relative survival ratios of the frozen-thawed erythrocytes by 10% and 20% (p<0.001), respectively. The SMFs had no effect on erythrocyte morphology and metabolite levels. However, membrane fluidity of the samples exposed to 0.8-T SMF decreased significantly (p<0.05) in the hydrophobic regions. For the dehydration stability experiments, the samples exposed to 0.8-T SMF exhibited significantly lower (p<0.05) hemolysis. These results demonstrate that a 0.8-T SMF decreases membrane fluidity and enhances erythrocyte membrane stability to resist dehydration damage caused by slow cooling procedures.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioUltra, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)