Merck
  • Crystallization and preliminary X-ray crystallographic analysis of the human CKIP-1 pleckstrin homology domain.
Crystallization and preliminary X-ray crystallographic analysis of the human CKIP-1 pleckstrin homology domain.
Acta crystallographica. Section F, Structural biology and crystallization communications (2013-03-23)
Ping Li, Yuli Xu, Xin Li, Mark Bartlam
ABSTRACT
The casein kinase 2 interacting protein-1 (CKIP-1) is involved in many cellular functions, including apoptosis, signalling pathways, cell growth, cytoskeleton and bone formation. Its N-terminal pleckstrin homology (PH) domain is thought to play an important role in membrane localization and controls shuttling of CKIP-1 between the plasma membrane and nucleus. In this study, the human CKIP-1 PH domain was purified but problems were encountered with nucleic acid contamination. An S84D/S86D/S88D triple mutant designed to abolish nucleic acid binding was purified and successfully crystallized. Single crystals diffracted to 1.7 Å resolution and belonged to space group P4₃2₁2 with unit-cell parameters a=53.0, b=53.0, c=113.8 Å, α=β=γ=90.0°.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)