Merck
Catalytic DNA with phosphatase activity.
Proceedings of the National Academy of Sciences of the United States of America (2013-03-20)
Jagadeeswaran Chandrasekar, Scott K Silverman
ABSTRACT
Catalytic DNA sequences (deoxyribozymes, DNA enzymes, or DNAzymes) have been identified by in vitro selection for various catalytic activities. Expanding the limits of DNA catalysis is an important fundamental objective and may facilitate practical utility of catalysts that can be obtained from entirely unbiased (random) sequence populations. In this study, we show that DNA can catalyze Zn(2+)-dependent phosphomonoester hydrolysis of tyrosine and serine side chains (i.e., exhibit phosphatase activity). The best deoxyribozyme decreases the half-life for phosphoserine hydrolysis from as high as >10(10) y to <1 h. The phosphatase activity also occurs with nonpeptidic substrates but with reduced efficiency, indicating a preference for phosphopeptides. The newly identified deoxyribozymes can function with multiple turnover using free peptide substrates, have activity in the presence of human cell lysate or BSA, and catalyze dephosphorylation of a larger protein substrate, suggesting broader application of DNA catalysts as artificial phosphatases.
Sigma-Aldrich
Malachite Green solution, for microscopy
Sigma-Aldrich
O-Phospho-DL-serine, ≥98.0% (NT)
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green oxalate salt, certified by the BSC
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade