Merck
  • Transcriptional regulation of the ADP-glucose pyrophosphorylase isoforms in the leaf and the stem under long and short photoperiod in lentil.
Transcriptional regulation of the ADP-glucose pyrophosphorylase isoforms in the leaf and the stem under long and short photoperiod in lentil.
Plant science : an international journal of experimental plant biology (2013-03-19)
Ayse Bengisu Seferoglu, Ibrahim Baris, Hande Morgil, Isil Tulum, Sule Ozdas, Gul Cevahir, Ibrahim Halil Kavakli
ABSTRACT
ADP-glucose pyrophosphorylase (AGPase) is a key enzyme in plant starch biosynthesis. It contains large (LS) and small (SS) subunits encoded by two different genes. In this study, we explored the transcriptional regulation of both the LS and SS subunits of AGPase in stem and leaf under different photoperiods length in lentil. To this end, we first isolated and characterized different isoforms of the LS and SS of lentil AGPase and then we performed quantitative real time PCR (qPCR) to see the effect of photoperiod length on the transcription of the AGPase isforms under the different photoperiod regimes in lentil. Analysis of the qPCR results revealed that the transcription of different isoforms of the LSs and the SSs of lentil AGPase are differentially regulated when photoperiod shifted from long-day to short-day in stem and leaves. While transcript levels of LS1 and SS2 in leaf significantly decreased, overall transcript levels of SS1 increased in short-day regime. Our results indicated that day length affects the transcription of lentil AGPase isoforms differentially in stems and leaves most likely to supply carbon from the stem to other tissues to regulate carbon metabolism under short-day conditions.
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble