Merck
  • Adsorption of lactic acid on chiral Pt surfaces--a density functional theory study.
Adsorption of lactic acid on chiral Pt surfaces--a density functional theory study.
The Journal of chemical physics (2013-03-08)
J-H Franke, D S Kosov
ABSTRACT
The adsorption of the chiral molecule lactic acid on chiral Pt surfaces is studied by density functional theory calculations. First, we study the adsorption of L-lactic acid on the flat Pt(111) surface. Using the optimed PBE - van der Waals (oPBE-vdW) functional, which includes van der Waals forces on an ab initio level, it is shown that the molecule has two binding sites, a carboxyl and the hydroxyl oxygen atoms. Since real chiral surfaces are (i) known to undergo thermal roughening that alters the distribution of kinks and step edges but not the overall chirality and (ii) kink sites and edge sites are usually the energetically most favored adsorption sites, we focus on two surfaces that allow qualitative sampling of the most probable adsorption sites. We hereby consider chiral surfaces exhibiting (111) facets, in particular, Pt(321) and Pt(643). The binding sites are either both on kink sites-which is the case for Pt(321) or on one kink site-as on Pt(643). The binding energy of the molecule on the chiral surfaces is much higher than on the Pt(111) surface. We show that the carboxyl group interacts more strongly than the hydroxyl group with the kink sites. The results indicate the possible existence of very small chiral selectivities of the order of 20 meV for the Pt(321) and Pt(643) surfaces. L-lactic acid is more stable on Pt(321)(S) than D-lactic acid, while the chiral selectivity is inverted on Pt(643)(S). The most stable adsorption configurations of L- and D-lactic acid are similar for Pt(321) but differ for Pt(643). We explore the impact of the different adsorption geometries on the work function, which is important for field ion microscopy.
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
Sigma-Aldrich
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.10 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, sponge, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, thermocouple grade
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis